(3-Chloro-2-fluoro-5-methylphenyl)(ethyl)sulphane ≥95%

Supplier: AOBChem USA

Warning

327316-5G 327316-25G 327316-500MG 327316-1G
77632-750EA 1441 USD
77632-750 77655-074 77646-088 77646-086
(3-Chloro-2-fluoro-5-methylphenyl)(ethyl)sulphane ≥95%
(3-Chloro-2-fluoro-5-methylphenyl)(ethyl)sulphane

Precaución: For Research Use Only

Formula: C₉H₁₀ClFS
Storage Temperature: Ambient
MDL Number: MFCD35179383
CAS Number: 3002542-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR