5-Bromo-2-hydroxy-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32729-5G 32729-1G 32729-25G 32729-500MG
77632-748EA 871 USD
77632-748 77638-098 77655-072 77646-084
5-Bromo-2-hydroxy-3-methylbenzyl alcohol ≥97%
5-Bromo-2-hydroxy-3-methylbenzyl alcohol

Precaución: For Research Use Only

Formula: C₈H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD16249637
CAS Number: 77691-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR