1-(3-Bromo-6-chloro-2-fluorophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

327150-5G 327150-1G 327150-500MG
77628-612EA 1484.23 USD
77628-612 77638-096 77646-082
1-(3-Bromo-6-chloro-2-fluorophenyl)propan-1-ol ≥95%
1-(3-Bromo-6-chloro-2-fluorophenyl)propan-1-ol

Precaución: For Research Use Only

Formula: C₉H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD35179219
CAS Number: 943830-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR