1-(3-Bromo-6-chloro-2-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

327149-5G 327149-1G 327149-25G 327149-500MG
77632-744EA 1326 USD
77632-744 77638-094 77655-068 77646-080
1-(3-Bromo-6-chloro-2-fluorophenyl)ethanol ≥95%
1-(3-Bromo-6-chloro-2-fluorophenyl)ethanol

Precaución: For Research Use Only

Formula: C₈H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD29913449
CAS Number: 943830-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR