2-(5-Bromopyridin-2-yl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

32693-5G 32693-25G
77634-786EA 131 USD
77634-786 77658-748
2-(5-Bromopyridin-2-yl)propan-2-ol ≥97%
2-(5-Bromopyridin-2-yl)propan-2-ol

Precaución: For Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11865230
CAS Number: 290307-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR