5-Chloro-2-ethoxypyridine-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

32683-5G 32683-1G 32683-25G 32683-500MG
77632-742EA 1198 USD
77632-742 77638-090 77658-746 77646-076
5-Chloro-2-ethoxypyridine-4-carboxylic acid ≥97%
5-Chloro-2-ethoxypyridine-4-carboxylic acid

Precaución: For Research Use Only

Formula: 201.607 g/mol
MW: C₈H₈ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD30529848
CAS Number: 1593360-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR