4-Bromo-3-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32620-5G 32620-500MG 32620-25G 32620-1G
77632-738EA 1289 USD
77632-738 77652-662 77658-744 77642-074
4-Bromo-3-formylphenylboronic acid pinacol ester ≥97%
4-Bromo-3-formylphenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: 310.979 g/mol
MW: C₁₃H₁₆BBrO₃
Storage Temperature: Ambient
MDL Number: MFCD28147940
CAS Number: 1417200-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR