(Z)-1-(p-Tolyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

325472-5G 325472-1G 325472-500MG
77628-604EA 1484.23 USD
77628-604 77638-084 77650-096
(Z)-1-(p-Tolyl)but-2-en-1-ol ≥95%
(Z)-1-(p-Tolyl)but-2-en-1-ol

Precaución: For Research Use Only

Formula: 162.228 g/mol
MW: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD35178642
CAS Number: 124745-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR