4-Bromo-4'-methylbenzophenone ≥97%

Supplier: AOBChem USA

Warning

325471-5G 325471-25G 325471-1G 325471-500MG
77634-784EA 342 USD
77634-784 77655-060 77642-070 77646-070
4-Bromo-4'-methylbenzophenone ≥97%
4-Bromo-4'-methylbenzophenone

Precaución: For Research Use Only

Formula: C₁₄H₁₁BrO
MW: 275.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00625706
CAS Number: 76693-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR