(3-Bromonaphthalen-2-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

325364-5G 325364-25G
77634-782EA 160 USD
77634-782 77658-742
(3-Bromonaphthalen-2-yl)boronic acid ≥97%
(3-Bromonaphthalen-2-yl)boronic acid

Precaución: For Research Use Only

Formula: C₁₀H₈BBrO₂
MW: 250.8843 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18410706
CAS Number: 1301205-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR