1-Bromo-2-ethoxy-3-fluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

32525-5G 32525-1G 32525-25G 32525-500MG
77634-780EA 576 USD
77634-780 77638-082 77658-740 77650-094
1-Bromo-2-ethoxy-3-fluoro-4-methylbenzene ≥97%
1-Bromo-2-ethoxy-3-fluoro-4-methylbenzene

Precaución: For Research Use Only

Formula: 233.078 g/mol
MW: C₉H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD32631802
CAS Number: 1871732-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR