3-Fluoro-2-hydroxy-6-methylanisole ≥95%

Supplier: AOBChem USA

Warning

32461-5G 32461-500MG 32461-25G 32461-1G
77632-736EA 1380 USD
77632-736 77652-660 77655-058 77642-068
3-Fluoro-2-hydroxy-6-methylanisole ≥95%
3-Fluoro-2-hydroxy-6-methylanisole

Precaución: For Research Use Only

Formula: C₈H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD22415073
CAS Number: 1780657-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR