Methyl 2-chloro-5-fluoro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

32446-5G 32446-25G 32446-1G 32446-500MG
77632-734EA 1198 USD
77632-734 77655-056 77642-066 77650-092
Methyl 2-chloro-5-fluoro-3-(trifluoromethyl)benzoate ≥97%
Methyl 2-chloro-5-fluoro-3-(trifluoromethyl)benzoate

Precaución: For Research Use Only

Formula: 256.581 g/mol
MW: C₉H₅ClF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD28752630
CAS Number: 1805955-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR