2-Chloro-3-methoxybenzoic acid methyl ester ≥95%

Supplier: AOBChem USA

Warning

32437-5G 32437-25G 32437-1G 32437-500MG
77634-778EA 572 USD
77634-778 77655-054 77642-064 77650-090
2-Chloro-3-methoxybenzoic acid methyl ester ≥95%
2-Chloro-3-methoxybenzoic acid methyl ester

Precaución: For Research Use Only

Formula: C₉H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD10566213
CAS Number: 59425-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR