[3-Bromo-2-(methylsulfanyl)phenyl]methanol ≥95%

Supplier: AOBChem USA

Warning

32433-5G 32433-1G 32433-25G 32433-500MG
77632-730EA 1117 USD
77632-730 77638-078 77655-052 77646-068
[3-Bromo-2-(methylsulfanyl)phenyl]methanol ≥95%
[3-Bromo-2-(methylsulphanyl)phenyl]methanol

Precaución: For Research Use Only

Formula: 233.125 g/mol
MW: C₈H₉BrOS
Storage Temperature: Ambient
MDL Number: MFCD30480187
CAS Number: 2008218-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR