2,4-Dimethoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32417-5G 32417-25G 32417-1G 32417-500MG
77632-728EA 734 USD
77632-728 77658-734 77642-062 77650-088
2,4-Dimethoxy-5-methylbenzoic acid ≥97%
2,4-Dimethoxy-5-methylbenzoic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD04038816
CAS Number: 50625-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR