(2-Chloro-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

32398-5G 32398-25G 32398-1G 32398-500MG
77632-726EA 918 USD
77632-726 77655-044 77642-060 77650-084
(2-Chloro-3-methoxyphenyl)methanol ≥97%
(2-Chloro-3-methoxyphenyl)methanol

Precaución: For Research Use Only

Formula: 172.609 g/mol
MW: C₈H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD18393574
CAS Number: 52516-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR