2-(4-Bromo-1-fluoronaphthalen-2-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

323901-5G 323901-1G 323901-25G 323901-500MG
77628-602EA 1484.23 USD
77628-602 77638-074 77655-040 77650-082
2-(4-Bromo-1-fluoronaphthalen-2-yl)propan-2-ol ≥95%
2-(4-Bromo-1-fluoronaphthalen-2-yl)propan-2-ol

Precaución: For Research Use Only

Formula: 283.136 g/mol
MW: C₁₃H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD35178179
CAS Number: 3002508-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR