1-(4-Fluoronaphthalen-1-yl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

323890-1G 323890-500MG 323890-5G
77638-072EA 676 USD
77638-072 77652-656 77655-038
1-(4-Fluoronaphthalen-1-yl)prop-2-en-1-ol ≥95%
1-(4-Fluoronaphthalen-1-yl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 202.224 g/mol
MW: C₁₃H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD35178169
CAS Number: 1430748-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR