2-(4-Fluoronaphthalen-1-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

323888-5G 323888-25G 323888-1G 323888-500MG
77632-724EA 1186 USD
77632-724 77655-036 77642-058 77650-080
2-(4-Fluoronaphthalen-1-yl)propan-2-ol ≥95%
2-(4-Fluoronaphthalen-1-yl)propan-2-ol

Precaución: For Research Use Only

Formula: 204.24 g/mol
MW: C₁₃H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD20528508
CAS Number: 55831-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR