Ethyl(4-fluoronaphthalen-1-yl)sulfane ≥95%

Supplier: AOBChem USA

Warning

323882-5G 323882-25G 323882-1G 323882-500MG
77632-722EA 989 USD
77632-722 77655-034 77642-056 77646-062
Ethyl(4-fluoronaphthalen-1-yl)sulfane ≥95%
Ethyl(4-fluoronaphthalen-1-yl)sulphane

Precaución: For Research Use Only

Formula: 206.279 g/mol
MW: C₁₂H₁₁FS
Storage Temperature: Ambient
MDL Number: MFCD35178165
CAS Number: 3002503-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR