2-(3-Chloro-4-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

323825-5G 323825-500MG 323825-25G 323825-1G
77632-720EA 1320 USD
77632-720 77652-654 77658-732 77642-054
2-(3-Chloro-4-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(3-Chloro-4-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 238.634 g/mol
MW: C₁₀H₁₀ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD35178110
CAS Number: 2229214-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR