1-(5-Bromobenzo[d][1,3]dioxol-4-yl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

323684-5G 323684-1G 323684-500MG 323684-25G
77628-600EA 1484.23 USD
77628-600 77638-070 77652-652 77655-030
1-(5-Bromobenzo[d][1,3]dioxol-4-yl)prop-2-en-1-ol ≥95%
1-(5-Bromobenzo[d][1,3]dioxol-4-yl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 257.081 g/mol
MW: C₁₀H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD35177987
CAS Number: 1391001-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR