(E)-4-(3-fluorophenyl)-4-oxobut-2-enoic acid ≥95%

Supplier: AOBChem USA

Warning

323581-5G 323581-25G 323581-1G 323581-500MG
77632-718EA 1357 USD
77632-718 77655-026 77642-050 77646-058
(E)-4-(3-fluorophenyl)-4-oxobut-2-enoic acid ≥95%
(E)-4-(3-fluorophenyl)-4-oxobut-2-enoic acid

Precaución: For Research Use Only

Formula: 194.1591 g/mol
MW: C₁₀H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD20645616
CAS Number: 191014-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR