Ethyl 2-chloro-6-ethoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

32352-5G 32352-25G 32352-1G
77634-774EA 243 USD
77634-774 77655-024 77642-048
Ethyl 2-chloro-6-ethoxynicotinate ≥97%
Ethyl 2-chloro-6-ethoxynicotinate

Precaución: For Research Use Only

Formula: 229.66 g/mol
MW: C₁₀H₁₂ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD30529809
CAS Number: 1003578-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR