(5-Chloro-2,4-dimethoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

32340-5G 32340-25G 32340-1G 32340-500MG
77632-714EA 1259 USD
77632-714 77655-020 77642-044 77650-074
(5-Chloro-2,4-dimethoxyphenyl)methanol ≥95%
(5-Chloro-2,4-dimethoxyphenyl)methanol

Precaución: For Research Use Only

Formula: 202.635 g/mol
MW: C₉H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD28060084
CAS Number: 943109-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR