5-Chloro-2,4-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32335-5G 32335-1G 32335-25G 32335-500MG
77632-712EA 703 USD
77632-712 77638-066 77658-730 77646-054
5-Chloro-2,4-dimethoxybenzoic acid ≥97%
5-Chloro-2,4-dimethoxybenzoic acid

Precaución: For Research Use Only

Formula: 216.618 g/mol
MW: C₉H₉ClO₄
Storage Temperature: Ambient
MDL Number: MFCD13704128
CAS Number: 23053-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR