2-Chloro-4-morpholinobenzenecarboxylic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-morpholinobenzoic acid

323315-5G 323315-25G 323315-1G 323315-500MG
77634-772EA 597 USD
77634-772 77658-728 77642-042 77650-072
2-Chloro-4-morpholinobenzenecarboxylic acid ≥95%
2-Chloro-4-morpholinobenzenecarboxylic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₂ClNO₃
MW: 241.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425732
CAS Number: 175153-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR