5-Bromo-2,4-dimethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32327-5G 32327-1G 32327-500MG
77655-018EA 1989 USD
77655-018 77642-038 77650-068
5-Bromo-2,4-dimethoxyphenylboronic acid ≥97%
5-Bromo-2,4-dimethoxyphenylboronic acid

Precaución: For Research Use Only

Formula: C₈H₁₀BBrO₄
Storage Temperature: Ambient
MDL Number: MFCD31561302
CAS Number: 2414154-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR