5-Bromo-2-trifluoromethoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32317-5G 32317-25G 32317-1G 32317-500MG
77634-770EA 258 USD
77634-770 77658-726 77642-034 77646-050
5-Bromo-2-trifluoromethoxyphenylboronic acid pinacol ester ≥97%
5-Bromo-2-trifluoromethoxyphenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: 366.967 g/mol
MW: C₁₃H₁₅BBrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD30480185
CAS Number: 2121515-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR