1-[4-(Ethylthio)phenyl]pyrrolidine ≥95%

Supplier: AOBChem USA

Warning

323160-5G 323160-1G 323160-25G 323160-500MG
77628-590EA 1484.23 USD
77628-590 77638-064 77655-016 77650-066
1-[4-(Ethylthio)phenyl]pyrrolidine ≥95%
1-[4-(Ethylthio)phenyl]pyrrolidine

Precaución: For Research Use Only

Formula: 207.34 g/mol
MW: C₁₂H₁₇NS
Storage Temperature: Ambient
MDL Number: MFCD35177716
CAS Number: 2243730-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR