5-Phenoxypyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

32264-5G 32264-500MG 32264-25G 32264-1G
77632-710EA 1191 USD
77632-710 77652-648 77655-012 77642-030
5-Phenoxypyridine-3-carboxylic acid ≥97%
5-Phenoxypyridine-3-carboxylic acid

Precaución: For Research Use Only

Formula: 215.205 g/mol
MW: C₁₂H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD20038718
CAS Number: 668969-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR