Methyl 3-chloro-2-fluoro-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

32260-5G 32260-1G 32260-25G 32260-500MG
77632-708EA 1117 USD
77632-708 77638-058 77655-010 77650-064
Methyl 3-chloro-2-fluoro-4-methoxybenzoate ≥97%
Methyl 3-chloro-2-fluoro-4-methoxybenzoate

Precaución: For Research Use Only

Formula: 218.609 g/mol
MW: C₉H₈ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD30737493
CAS Number: 2121309-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR