(3-Chloro-2-fluoro-4-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

32258-5G 32258-1G 32258-500MG
77628-586EA 1484.23 USD
77628-586 77642-028 77646-044
(3-Chloro-2-fluoro-4-methoxyphenyl)methanol ≥97%
(3-Chloro-2-fluoro-4-methoxyphenyl)methanol

Precaución: For Research Use Only

Formula: 190.599 g/mol
MW: C₈H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28523088
CAS Number: 1616601-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR