3-Chloro-2-fluoro-4-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32255-5G 32255-25G 32255-1G 32255-500MG
77632-706EA 1198 USD
77632-706 77655-008 77642-024 77650-062
3-Chloro-2-fluoro-4-methoxyphenylboronic acid pinacol ester ≥97%
3-Chloro-2-fluoro-4-methoxyphenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₃H₁₇BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD30737492
CAS Number: 2121512-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR