2'-Methyl-4'-nitroacetanilide ≥95%

Supplier: AOBChem USA

Warning

321980-5G 321980-25G
77634-758EA 86 USD
77634-758 77658-714
2'-Methyl-4'-nitroacetanilide ≥95%
2'-Methyl-4'-nitroacetanilide

Precaución: For Research Use Only

Formula: C₉H₁₀N₂O₃
MW: 194.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00033896
CAS Number: 2719-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR