4'-Fluoro-3'-nitroacetanilide ≥97%

Supplier: AOBChem USA

Warning

321966-5G 321966-25G
77634-756EA 59 USD
77634-756 77658-712
4'-Fluoro-3'-nitroacetanilide ≥97%
4'-Fluoro-3'-nitroacetanilide

Precaución: For Research Use Only

Formula: C₈H₇FN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00585083
CAS Number: 351-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR