Ethyl 4-(4-chlorophenyl)-2-oxo-3-butynoate ≥95%

Supplier: AOBChem USA

Warning

321645-5G 321645-1G 321645-25G 321645-500MG
77632-702EA 828 USD
77632-702 77638-052 77655-000 77646-032
Ethyl 4-(4-chlorophenyl)-2-oxo-3-butynoate ≥95%
Ethyl 4-(4-chlorophenyl)-2-oxo-3-butynoate

Precaución: For Research Use Only

Formula: 236.65 g/mol
MW: C₁₂H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD32856707
CAS Number: 158555-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR