3-(4-Chlorophenyl)-1-phenyl-2-propyn-1-one ≥95%

Supplier: AOBChem USA

Warning

321632-5G 321632-25G 321632-1G 321632-500MG
77632-700EA 796 USD
77632-700 77654-998 77642-016 77646-030
3-(4-Chlorophenyl)-1-phenyl-2-propyn-1-one ≥95%
3-(4-Chlorophenyl)-1-phenyl-2-propyn-1-one

Precaución: For Research Use Only

Formula: C₁₅H₉ClO
Storage Temperature: Ambient
MDL Number: MFCD32856815
CAS Number: 29776-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR