4-(4-Chlorophenyl)-3-butyn-2-one ≥95%

Supplier: AOBChem USA

Warning

321630-5G 321630-25G 321630-1G 321630-500MG
77632-698EA 1168 USD
77632-698 77654-996 77642-014 77646-028
4-(4-Chlorophenyl)-3-butyn-2-one ≥95%
4-(4-Chlorophenyl)-3-butyn-2-one

Precaución: For Research Use Only

Formula: C₁₀H₇ClO
MW: 178.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24689111
CAS Number: 39180-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR