1-(2-Iodoethynyl)-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

321089-1G 321089-5G 321089-500MG
77638-050EA 656 USD
77638-050 77654-994 77650-056
1-(2-Iodoethynyl)-4-methylbenzene ≥95%
1-(2-Iodoethynyl)-4-methylbenzene

Precaución: For Research Use Only

Formula: C₉H₇I
Storage Temperature: Ambient
MDL Number: MFCD22666213
CAS Number: 33675-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR