1-(3-Bromo-5-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

32065-5G 32065-25G 32065-1G 32065-500MG
77628-582EA 1484.23 USD
77628-582 77635-640 77638-046 77650-052
1-(3-Bromo-5-fluorophenyl)ethanol ≥95%
1-(3-Bromo-5-fluorophenyl)ethanol

Precaución: For Research Use Only

Formula: 219.051 g/mol
MW: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD21773565
CAS Number: 627527-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR