1-(4-Methoxy-2-(trifluoromethyl)phenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

320454-5G 320454-1G 320454-500MG
77628-580EA 1777.78 USD
77628-580 77642-010 77646-026
1-(4-Methoxy-2-(trifluoromethyl)phenyl)prop-2-en-1-ol ≥95%
1-(4-Methoxy-2-(trifluoromethyl)phenyl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 232.199 g/mol
MW: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD30488652
CAS Number: 2060040-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR