3-(4-Methoxy-2-(trifluoromethyl)phenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

320452-5G 320452-500MG 320452-25G 320452-1G
77628-578EA 1513.07 USD
77628-578 77652-644 77658-702 77642-008
3-(4-Methoxy-2-(trifluoromethyl)phenyl)oxetan-3-ol ≥95%
3-(4-Methoxy-2-(trifluoromethyl)phenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: 248.198 g/mol
MW: C₁₁H₁₁F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD35176850
CAS Number: 3002523-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR