Cyclopropyl(4-methoxy-2-(trifluoromethyl)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

320451-1G 320451-5G 320451-500MG
77638-044EA 628 USD
77638-044 77654-992 77646-024
Cyclopropyl(4-methoxy-2-(trifluoromethyl)phenyl)methanol ≥95%
Cyclopropyl(4-methoxy-2-(trifluoromethyl)phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD35176849
CAS Number: 2353906-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR