Ethyl(4-methoxy-2-(trifluoromethyl)phenyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
320447-5G
320447-25G
320447-1G
320447-500MG
77628-576EA
1484.23
USD
77628-576
77635-638
77638-042
77652-642
Ethyl(4-methoxy-2-(trifluoromethyl)phenyl)sulfane ≥95%
Ethyl(4-methoxy-2-(trifluoromethyl)phenyl)sulphane
Precaución: For Research Use Only
Formula:
236.254 g/mol MW: C₁₀H₁₁F₃OS Storage Temperature: Ambient |
MDL Number:
MFCD35176846 CAS Number: 3002523-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...