2-(4-Methoxy-2-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

320446-5G 320446-1G 320446-500MG
77628-574EA 1724.22 USD
77628-574 77642-006 77646-022
2-(4-Methoxy-2-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(4-Methoxy-2-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 234.215 g/mol
MW: C₁₁H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD30488480
CAS Number: 1785147-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR