2-(Tert-butylthio)-4-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

320381-5G 320381-1G 320381-25G 320381-500MG
77628-572EA 1484.23 USD
77628-572 77638-038 77654-988 77650-050
2-(Tert-butylthio)-4-fluorobenzaldehyde ≥97%
2-(Tert-butylthio)-4-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: 212.284 g/mol
MW: C₁₁H₁₃FOS
Storage Temperature: Ambient
MDL Number: MFCD35176789
CAS Number: 3002538-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR