(2,4-Bis(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥97%
Supplier: AOBChem USA
|
Warning
|
320370-5G
320370-25G
320370-1G
320370-500MG
77632-694EA
1441
USD
77632-694
77654-986
77642-004
77650-048
(2,4-Bis(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥97%
(2,4-Bis(trifluoromethyl)phenyl)(cyclopropyl)methanol
Precaución: For Research Use Only
Formula:
C₁₂H₁₀F₆O Storage Temperature: Ambient |
MDL Number:
MFCD35176778 CAS Number: 2377755-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...