(6-Bromo-5-methoxypyridin-3-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

32017-5G 32017-1G 32017-25G 32017-500MG
77632-692EA 1408 USD
77632-692 77638-036 77658-700 77646-018
(6-Bromo-5-methoxypyridin-3-yl)methanol ≥95%
(6-Bromo-5-methoxypyridin-3-yl)methanol

Precaución: For Research Use Only

Formula: 218.048 g/mol
MW: C₇H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD28741111
CAS Number: 1805577-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR